ContactUsLinkComponent
VWR®, Microcentrifuge tubes, 1,5 ml
VWR®, Microcentrifuge tubes, 1,5 ml
  VWRU89000-028
   Low Manufacturing Impact
   Sustainable Packaging
undefined
VWR®, Microcentrifuge tubes, 1,5 ml
  VWRU89000-028
   Low Manufacturing Impact
   Sustainable Packaging
  • Type:
    with attached lid
  • Version:
    graduated, conical bottom, non sterile
  • Capacity:
    1,5 ml
  • Colour:
    Natural
  • Seq:
    03
  • Environmentally Preferable:
    Low Manufacturing Impact,
    Sustainable Packaging
  • Pk:
    500
PP, disposable, graduated.
  • RNase- and DNase-free
  • Autoclavable
  • Tubes can be centrifuged to 20000×g
Tubes feature flat caps and frosted areas on the sides for easy labelling. Snap caps can be penetrated by a syringe. Easy-to-open caps will not pop open during centrifuging.
 : Manufactured in a Class 100000 cleanroom environment. Manufactured under ISO 13485:2016 and ISO 9001:2015 quality management systems. Meet the requirements of RoHS.
 : All tubes packaged in resealable bags.