ContactUsLinkComponent
VWR®, Sterilisation Boxes for Pipettes
VWR®, Sterilisation Boxes for Pipettes
Catalog # VWRU82027-608
Supplier:  VWR Collection
undefined
VWR®, Sterilisation Boxes for Pipettes
Catalog # VWRU82027-608
Supplier:  VWR Collection
Specifications
  • Description:
    Square box for pipettes
  • Seq:
    03
  • W×D×H:
    51×51×419 mm
  • Pk:
    1
Specifications
About this item
Pipette sterilisation boxes are designed to secure and protect pipettes during the sterilisation process.
  • Designed to secure and protect pipettes during sterilisation processes
  • Stainless steel construction ensures high corrosion resistance
  • Multiple box styles accommodate a variety of pipette types