Order Entry
Austria
Orders LinkContactUsLinkComponent
Dual scale salinometer

Dual scale salinometer

1,000 - 1,250 g/ml ; 0 - 30 °Bé, 0,001 g/ml ; 0,1 °Bé, 195 mm
Catalog # ALLA1020PM125/20QP
Supplier:   Alla France
Dual scale salinometer
Catalog # ALLA1020PM125/20QP
Supplier:  Alla France
Dual scale salinometer
Dual scale salinometer

1,000 - 1,250 g/ml ; 0 - 30 °Bé, 0,001 g/ml ; 0,1 °Bé, 195 mm

Price
154,00 €
Log in for availability
1 * 5 ST
Product Details & Documents

This salinometer is designed for measurements of brine and salt.

Specifications
  • Length:
    195 mm
  • Division:
    0,001 g/ml ; 0,1 °Bé
  • Range:
    1,000 - 1,250 g/ml ; 0 - 30 °Bé