Order Entry
Austria
ContactUsLinkComponent
Copper(II) oxide, wire ACS (0.65x6 mm) for elementary analysis, Supelco®
Copper(II) oxide, wire ACS (0.65x6 mm) for elementary analysis, Supelco®
Catalog # 1.02767.0250
Supplier:  Merck
CAS Number:  
Copper(II) oxide, wire ACS (0.65x6 mm) for elementary analysis, Supelco®
Catalog # 1.02767.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g
  • Seq:
    3545
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (oxygen demand as CuO):
    ≥ 55.0 %
  • Chloride (Cl):
    ≤ 0.005 %
  • Total carbon (C):
    ≤ 0.002 %
  • Total nitrogen (N):
    ≤ 0.002 %
  • Total sulfur (as SO₄):
    ≤ 0.005 %

Specification Test Results

About this item

Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated
 :
 :79.55 g/mol
EINECS:215-269-1
UN:3077
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:1317-38-0
Formula:CuO
MDL Number:MFCD00010979
Boiling Pt:2000
Melting Pt:1326
ADR:9 III
Merck Index:13,02674
 :M-Clarity™ quality level = MQ100