Order Entry
Austria
Orders LinkContactUsLinkComponent
 

VWR®, Zero Air Generators

Supplier:   VWR Collection
VWR®, Zero Air Generators
Supplier:  VWR Collection
Certificates
VWR®, Zero Air Generators
VWR®, Zero Air Generators

2 Options Available Below

Product Details & Documents

Designed as a compact, hazard free, lower cost alternative to the user of expensive high pressure gas cylinders in the laboratory. Replacing cylinders with a gas generator brings advantages : increased safety, greater convenience, reduced cost and higher purity gas for more accurate analysis.

  • Produces zero air from compressed air
  • Ensures a flat and consistent base line
  • Short payback period

Purity: <0,05 ppm THC.

Inlet pressure 2,8 to 8,6 bar.

Inlet/outlet port size: ¹/₄" NPT female.

Product Family Options

Product Information

  • Flow rate
    W×D×H
    Availability
    Price
  • 7 l/min
    280×330×304 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • 18 l/min
    280×330×304 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.