
Potassium hydroxide 0.1 N in ethanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5645.1000
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Titer (mol/l):0.0995-0.1005
- A light red color can occur in time due to product characteristics:Passes Test
Specification Test Results
About this item
:
:56.11 g/mol
ADR:3,II
:Management of Change (MOC) category = N
EINECS:215-181-3
UN:2924
:GHS02
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553