Order Entry
Austria
ContactUsLinkComponent
Sodium pyruvate ≥99.0% (by non aqueous titration), white powder, BAKER ANALYZED®, J.T.Baker®
Sodium pyruvate ≥99.0% (by non aqueous titration), white powder, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium pyruvate ≥99.0% (by non aqueous titration), white powder, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    1% max.
  • Assay (CH₃COCOONa) (by non-aqueous titration):
    ≥99.0 %
  • Free Pyruvic Acid (CH₃COCOOH):
    ≤1 %
  • Solubility:
    Passes Test

Specification Test Results

Related Information

Frequently Bought Together