ContactUsLinkComponent
VWR®, Micro Pipettes, Disposable
VWR®, Micro Pipettes, Disposable
Catalog # VWRU53432-728
Supplier:  VWR Collection
undefined
VWR®, Micro Pipettes, Disposable
Catalog # VWRU53432-728
Supplier:  VWR Collection
Specifications
  • Colour code:
    Orange
  • Capacity:
    10 µl
  • Seq:
    02
  • Pk:
    250
Specifications
About this item
Borosilicate glass.
  • Calibrated to contain
  • With permanent fill lines
  • Colour-coded
Packaging: Packed in easy dispensing canisters of 250 micropipettes, plus one 38,1 cm aspirator tube. Each canister is marked with bold size designations for quick identification in the laboratory.