Spill control accumulation centers and ramps, EcoPolyBlend™
EcoPolyBlend™ accumulation centers offer high value secondary containment and are environmentally friendly-made of recycled, durable polyethylene. These handy platforms offer leak protection from drums, pails, batteries, or any other container of hazardous or non-hazardous liquids.
- Sturdy, removable grates make sump cleaning easy while low profile offers convenience when loading/unloading
- Load and unload heavy drum with ease
- Anti-skid ribs on the ramp provides maximum safety
Frequently Bought Together