Orders LinkContactUsLinkComponent
Filter papers for breweries
Filter papers for breweries
Filter papers for breweries
Anbieter:  Ahlstrom
Certificates
Filter papers for breweries
12 Options Available Below
Product Details & Documents
Ahlstrom-Munksjö offers filters for malt analysis and malt filtration according to MEBAK, ASBC and EBC standards.
Grade 41b: Medium fast filtering. Recommended for malt analysis.
Grade 15: Plain, for final attenuation of congress wort.
Grade 470: Plain kieselguhr paper used for fine turbidity.
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • BINZ120047
  • BINZ120048
  • BINZ120001
  • BINZ120049
  • BINZ120050
  • BINZ120051
  • BINZ120052
  • BINZ120053
  • BINZ120076
  • BINZ120054
  • BINZ146279
  • BINZ146282
  • Grade:
  • 15
  • 15
  • 15
  • 15
  • 15
  • 15
  • 15
  • 15
  • 15
  • 15
  • 41b
  • 41b
  • Weight:
  • 68 g/m²
  • 68 g/m²
  • 68 g/m²
  • 68 g/m²
  • 68 g/m²
  • 68 g/m²
  • 68 g/m²
  • 68 g/m²
  • 68 g/m²
  • 68 g/m²
  • 75 g/m²
  • 75 g/m²
  • Filtration speed:
  • 25 s/10 ml
  • 25 s/10 ml
  • 25 s/10 ml
  • 25 s/10 ml
  • 25 s/10 ml
  • 25 s/10 ml
  • 25 s/10 ml
  • 25 s/10 ml
  • 25 s/10 ml
  • 25 s/10 ml
  • 22 s/10 ml
  • 22 s/10 ml
Specifications
Product Family Options
Circles
Product Information
  • Grade
    Size
    Verfügbarkeit
    Preis
  • 15
    55 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 6,70 €
    6,70 €
    1 * 100 ST
       6,70 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    BINZ120047
    Grade
    15
    Weight
    68 g/m²
    Filtration speed
    25 s/10 ml
  • 15
    70 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 7,10 €
    7,10 €
    1 * 100 ST
       7,10 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    BINZ120048
    Grade
    15
    Weight
    68 g/m²
    Filtration speed
    25 s/10 ml
  • 15
    90 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 6,90 €
    6,90 €
    1 * 100 ST
       6,90 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    BINZ120001
    Grade
    15
    Weight
    68 g/m²
    Filtration speed
    25 s/10 ml
  • 15
    110 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 10,30 €
    10,30 €
    1 * 100 ST
       10,30 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    BINZ120049
    Grade
    15
    Weight
    68 g/m²
    Filtration speed
    25 s/10 ml
  • 15
    125 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 12,00 €
    12,00 €
    1 * 100 ST
       12,00 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    BINZ120050
    Grade
    15
    Weight
    68 g/m²
    Filtration speed
    25 s/10 ml
  • 15
    150 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,60 €
    16,60 €
    1 * 100 ST
       16,60 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    BINZ120051
    Grade
    15
    Weight
    68 g/m²
    Filtration speed
    25 s/10 ml
  • 15
    185 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,50 €
    16,50 €
    1 * 100 ST
       16,50 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    BINZ120052
    Grade
    15
    Weight
    68 g/m²
    Filtration speed
    25 s/10 ml
  • 15
    240 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 39,60 €
    39,60 €
    1 * 100 ST
       39,60 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    BINZ120053
    Grade
    15
    Weight
    68 g/m²
    Filtration speed
    25 s/10 ml
  • 15
    270 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 47,70 €
    47,70 €
    1 * 100 ST
       47,70 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    BINZ120076
    Grade
    15
    Weight
    68 g/m²
    Filtration speed
    25 s/10 ml
  • 15
    320 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 65,20 €
    65,20 €
    1 * 100 ST
       65,20 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    BINZ120054
    Grade
    15
    Weight
    68 g/m²
    Filtration speed
    25 s/10 ml
Folded filters
Product Information
  • Grade
    Size
    Verfügbarkeit
    Preis
  • 41b
    240 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 45,00 €
    45,00 €
    1 * 100 ST
       45,00 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    BINZ146279
    Grade
    41b
    Weight
    75 g/m²
    Filtration speed
    22 s/10 ml
  • 41b
    320 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 58,50 €
    58,50 €
    1 * 100 ST
       58,50 €
    Specifications:
    Mehr
    Specifications
    Cat. No.
    BINZ146282
    Grade
    41b
    Weight
    75 g/m²
    Filtration speed
    22 s/10 ml