Orders LinkContactUsLinkComponent
Microscope slides with printed rings
Microscope slides with printed rings
Microscopy slide with printed rings
Catalog # BELC5638-01920
Marketsource
Supplier:   BELLCO GLASS
Microscope slides with printed rings
Catalog # BELC5638-01920
Supplier:  BELLCO GLASS
Marketsource
Microscope slides with printed rings
Microscope slides with printed rings
Microscopy slide with printed rings
Price
391,00 €
Log in for availability
1 * 144 ST
Product Details & Documents
These microscopy slides with three printed rings allow for counting bacteria by spreading a known volume of solution over a known area for the calculation of bacteria per sample.
Area (ring): 1 cm²
Specifications
  • Description:
    Microscopy slide with printed rings