Orders LinkContactUsLinkComponent
Precision thermometers
Precision thermometers
Supplier:   Ludwig Schneider
Precision thermometers
Supplier:  Ludwig Schneider
Certificates
Precision thermometers
Precision thermometers
6 Options Available Below
Product Details & Documents
Precision thermometer, round, white back capillary form, red filling, stem type.
  • With permanent graduation and lettering
  • With serial number and certificate (conformance and accuracy)
 : Single packed in plastic square shucks.
Product Family Options
Product Information
  • Range
    Division
    Length
    Filling
    Immersion depth
    Availability
    Price
  • −100...+50 °C
    1,0 °C
    380 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,90 €
    43,90 €
    1 * 1 ST
       43,90 €
  • −100...+50 °C
    1,0 °C
    530 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,30 €
    53,30 €
    1 * 1 ST
       53,30 €
  • −20...+150 °C
    1,0 °C
    380 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,40 €
    29,40 €
    1 * 1 ST
       29,40 €
  • −20...+150 °C
    1,0 °C
    530 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,90 €
    42,90 €
    1 * 1 ST
       42,90 €
  • −10...+250 °C
    1,0 °C
    430 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,30 €
    33,30 €
    1 * 1 ST
       33,30 €
  • −10...+250 °C
    1,0 °C
    580 mm
    Red
    Total
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,50 €
    44,50 €
    1 * 1 ST
       44,50 €