Orders LinkContactUsLinkComponent
Interfaces
Interfaces
Supplier:   HUBER
Interfaces
Supplier:  HUBER
Certificates
Interfaces
3 Options Available Below
Product Details & Documents
Com.G@te®. The interfaces are integrated RS232 (bi-directional), RS485 (bi-directional), ECS external control signal, volt free contact, AIF analogue interface 0/4 to 20 mA or 0 to 10 V (bi-directional). Connections comply with NAMUR standard.
Product Family Options
Product Information
  • Availability
    Price
  • HUBE31217
    Com.G@te®, internal for Petite Fleur®, Grande Fleur®, Unichiller® with Pilot ONE, Ministats®, CC®-300BX to CC®-906w
    Com.G@te®, internal for Petite Fleur®, Grande Fleur®, Unichiller® with Pilot ONE, Ministats®, CC®-300BX to CC®-906w
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 715,00 €
    715,00 €
    1 * 1 ST
       715,00 €
  • HUBE6915
    Com.G@te®, external for Unistats®, CC®-E to CC®-205B
    Com.G@te®, external for Unistats®, CC®-E to CC®-205B
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 715,00 €
    715,00 €
    1 * 1 ST
       715,00 €
  • HUBE10019
    Holder for Com.G@te® for Unistats® (bench top models)
    Holder for Com.G@te® for Unistats® (bench top models)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 160,00 €
    160,00 €
    1 * 1 ST
       160,00 €