Gateway, Profibus
Gateway 3E (external, for Pilot ONE), 90 – 260V, Profibus
Catalog # HUBE10503
Supplier: HUBER
Jump to:
Product Details & Documents
This gateway enables the connection of temperature control units to Profibus systems.
Offers a range of possibilities for data communication within SPS and process control systems.
: Includes a mounting bracket and connection cable.
Specifications
- Model:Profibus
- Description:Gateway 3E (external, for Pilot ONE), 90 – 260V