Capillary adaptable FID jet
Supplier: Restek
Certificates
About this item
These capillary adaptable FID replacement jets are engineered with a fluted tip to guide the capillary column into the jet.
- For Agilent 5890/6850/6890 GCs
- Available in standard or Siltek treated high-performance versions
Threads in standard version are specially coated for easy installation and removal, also special processing ensures the highest degree of cleanliness.
Int.Ø×L: 0,28×61,5 mm.
The high-performance versions are siltek treated which are extremely inert for use with active compounds.
Frequently Bought Together