Orders LinkContactUsLinkComponent
Sponge sticks
Sponge sticks
Supplier:  3M Food Safety
Certificates
Sponge sticks
4 Options Available Below
Product Details & Documents
3M™ Sponge-Stick in sample bag with 10 ml D/E neutralising broth uses simplified packaging to minimise handling and preparation, and reduces waste.
  • Stick-mounted sponge design allows the ability to access hard to reach areas without directly handling the sponge
  • Stick-mounted sponge design provides the ability to easily snap off the sponge from the handle after swabbing
  • Simplified packaging minimises handling and preparation and reduces waste
Better testing begins with better samples and better samples are obtained when you use the best collection tools. 3M™ Sponge-Stick allows sampling without directly handling the sponge. It makes it easier to reach into drains, pipes and around equipment. Biocide-free cellulose sponge maintains organism viability. These are convenient and ready-to-use with consistent, reliable results every time.
 : Supplied in wet or dry formats to fit many applications.
Product Family Options
Product Information
  • Availability
    Price
  • MMMPSSL100
    Media type: Without media
    Media type: Without media
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 307,00 €
    307,00 €
    1 * 100 ST
       307,00 €
  • MMMPSSL10NB
    Media type: Neutralising buffer
    Media type: Neutralising buffer
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 307,00 €
    307,00 €
    1 * 100 ST
       307,00 €
  • MMMPSSL10BPW
    Media type: Buffered peptone water broth
    Media type: Buffered peptone water broth
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 307,00 €
    307,00 €
    1 * 100 ST
       307,00 €
  • MMMPSSL10LET
    Media type: Letheen broth
    Media type: Letheen broth
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 307,00 €
    307,00 €
    1 * 100 ST
       307,00 €