Multi-gas detectors, GasAlert MicroClip XL
Supplier: BW TECHNOLOGIES
Certificates
About this item
The MicroClip XL provides protection from atmospheric gas hazards together with extended battery life, especially in cold weather for oxygen (O₂), hydrogen sulfide (H₂S), carbon monoxide (CO) and combustibles.
- Rechargeable battery - Survives harsh temperatures, punctures and 20-foot drops
- Flashing green IntelliFlash™ light for compliance at a glance
- Reflex™ Technology ensures sensor responsiveness for safety, uptime and worker confidence
- Simultaneously monitor for up to four measurements
- Battery runtime of 18 hours in normal temperatures; cold weather guarantee of at least 12 hour runtime for full warranty period
For simple management of your records, calibration and bump testing, the optional MicroDock II and IntelliDoX automatic test and calibration system and Fleet Manager II software are available as options.
Certifications: Class I, Div. 1, Gr. A, B, C, D; ATEX: X g II 1 G, Ex ia IIC T4 Ga; IECEx: Ex ia IIC T4 Ga - EMI/RFI: EMC 2004/108/EC - IP68.
Delivery: Complete with specified sensor(s), stainless steel alligator clip and concussion-proof housing, rechargeable battery, wall outlet charging adaptor, calibration/test cap and hose, manual and multi-language CD manual.
Specifications
- Catalog No:
- 151-0244
- 151-0243
- 151-0240
- 151-0246
- 151-0245
- 151-0242
- 151-0241
- Model:
- MicroClip XL
- MicroClip XL
- MicroClip XL
- MicroClip XL
- MicroClip XL
- MicroClip XL
- MicroClip XL
- Alarm:
- Audible (95 dB), visual, Buzzer
- Audible (95 dB), visual, Buzzer
- Audible (95 dB), visual, Buzzer
- Audible (95 dB), visual, Buzzer
- Audible (95 dB), visual, Buzzer
- Audible (95 dB), visual, Buzzer
- Audible (95 dB), visual, Buzzer
- Battery life:
- 18 heures, avec charge en moins de six heures
- 18 heures, avec charge en moins de six heures
- 18 heures, avec charge en moins de six heures
- 18 heures, avec charge en moins de six heures
- 18 heures, avec charge en moins de six heures
- 18 heures, avec charge en moins de six heures
- 18 heures, avec charge en moins de six heures
- W×D×H:
- 60×31×113 mm
- 60×31×113 mm
- 60×31×113 mm
- 60×31×113 mm
- 60×31×113 mm
- 60×31×113 mm
- 60×31×113 mm
- Weight:
- 190 g
- 190 g
- 190 g
- 190 g
- 190 g
- 190 g
- 190 g
- Temperature range:
- −20…+50 °C
- −20…+50 °C
- −20…+50 °C
- −20…+50 °C
- −20…+50 °C
- −20…+50 °C
- −20…+50 °C
Specifications
Frequently Bought Together