ContactUsLinkComponent
Dual stage ultra high purity gas regulators
Dual stage ultra high purity gas regulators
Supplier:  Restek
Certificates
undefined
Dual stage ultra high purity gas regulators
Supplier:  Restek
Specifications
  • Catalog No:
  • REST21667
  • REST21669
  • REST21668
  • Max. inlet pressure:
  • 206,8 bar
  • 206,8 bar
  • 206,8 bar
  • Outlet pressure:
  • 0,00 - 10,34 bar
  • 0,00 - 10,34 bar
  • 0,00 - 10,34 bar