Anti-MARCH11 Rabbit Polyclonal Antibody
Catalog # BOSSBS-9345R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Membrane-associated ring Finger (C3HC4)11, E3 ubiquitin protein ligase
- Antigen symbol:MARCH11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Gene ID:441061
- Antigen synonyms:Membrane associated ring finger C3HC4 11|March11|Membrane-associated RING-CH protein XI.|Membrane-associated RING finger protein 11|MARCH-XI|E3 ubiquitin-protein ligase MARCH11|MARHB_HUMAN
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
E3 ubiquitin-protein ligase that mediates polyubiquitination of CD4. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates. May play a role in ubuquitin-dependent protein sorting in developmenting spermatids.
IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: MARCH11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat