Specifications
- Antibody type:Primary
- Antigen name:Membrane-associated ring Finger (C3HC4) 7, E3 ubiquitin protein ligase
- Antigen symbol:MARCH7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:64844
- Antigen synonyms:AXOT|RNF177|AXO|MARCH-VII
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Axotrophin is a stem cell gene that encodes a protein which is involved in T lymphocyte regulation (especially in regulating the proliferation) and leukemia inhibitory factor (LIF) release. LIF is a neuropoietic cytokine that is important for stem cell regulation and thymocyte stimulation. Both Axotrophin and LIF are linked to transplantation intolerance. Axotrophin is also involved in corpus callosum differentiation and may play a role in glial cell line-derived neurotrophic factor (GDNF)-dependent sensory neuron survival in the substantia gelatinosa of the adult spinal cord. Axotrophin is primarily expressed in the hippocampus, cortex, purkinje and granule cells of the cerebellum.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: MARCH7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat