Specifications
- Antibody type:Primary
- Antigen name:Dipeptidylpeptidase 6
- Antigen symbol:DPP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:1804
- Antigen synonyms:Dipeptidylpeptidase VI|Dipeptidyl peptidase IV like protein|Dipeptidyl aminopeptidase related protein|DPPX|Dipeptidylpeptidase 6|DPP6_HUMAN.|Dipeptidyl aminopeptidase like protein 6|Dipeptidyl peptidase 6
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
DPP6 is a Type-II serine proteinase of the clan SC. The clan SC proteinases have a catalytic triad of Ser-Asp-His, and like other Serine proteinases, the active site serine is in a Gly-Xaa-Ser-Xaa -Gly orientation. DPP6 has an Asp instead of Ser in the catalytic site. DPP6 is a member of a broader family of dipeptidyl peptidases including DPP4, FAP/Seprase, DPP2, DPP8, DPP9, DPP10, which have differing substrate specificity and tissue localizations. The surface-bound DPP6 is a homodimer, and cleavage of in the stalk region releases a shed form of DPP6. The shed is the form found in serum. DPP6 has been found in highest abundance in the brain, but also in the kidney, liver and lung.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: DPP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat