Anti-TEAD2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-8212R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:TEA domain family member 2
- Antigen symbol:TEAD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:8463
- Antigen synonyms:ETF|TEF4|TEAD-2|TEF-4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: TEAD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit