Order Entry
Austria
ContactUsLinkComponent
Anti-CCNJ Rabbit Polyclonal Antibody
Anti-CCNJ Rabbit Polyclonal Antibody
Catalog # BOSSBS-7890R
Supplier:  Bioss
Anti-CCNJ Rabbit Polyclonal Antibody
Catalog # BOSSBS-7890R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin J
  • Antigen symbol:
    CCNJ
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    54619
  • Antigen synonyms:
    CyclinJ
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Cyclins are a family of proteins involved in the progression of cells through the cell cycle. Cyclins are so named because their concentration varies in a cyclical fashion during the cell cycle; they are produced or degraded as needed in order to drive the cell through the different stages of the cell cycle. Cyclin J has an RNA expression pattern that suggests a possible role in early embryogenesis.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CCNJ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat