Anti-EME1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-7861R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Essential meiotic structure-specific endonuclease 1
- Antigen symbol:EME1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:146956
- Antigen synonyms:MMS4L|SLX2A
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
EME1 complexes with methyl methanesulfonate-sensitive UV-sensitive 81 protein (MUS81) to form an endonuclease complex which cleaves branched DNA structures, especially those arising during stalled DNA replication. The protein may be involved in repairing DNA damage and in maintaining genomic stability. It interacts with specifc DNA structures including nicked Holliday junctions, 3'-flap structures and aberrant replication fork structures. Alternative splicing results in multiple transcript variants.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: EME1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat