Order Entry
Austria
ContactUsLinkComponent
Anti-MCHR1 Rabbit Polyclonal Antibody
Anti-MCHR1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-6626R
Supplier:  Bioss
undefined
Anti-MCHR1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-6626R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Melanin-Concentrating Hormone Receptor 1
  • Antigen symbol:
    MCHR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    liquid
  • Gene ID:
    2847
  • Antigen synonyms:
    MCH-1R|GPR24|SLC1|MCH1R|SLC-1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Immunogen:
    60-113/422
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Receptor for melanin-concentrating hormone, coupled to both G proteins that inhibit adenylyl cyclase and G proteins that activate phosphoinositide hydrolysis.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: MCHR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat