Order Entry
Austria
ContactUsLinkComponent
Anti-PRKD3 Rabbit Polyclonal Antibody
Anti-PRKD3 Rabbit Polyclonal Antibody
  BOSSBS-5564R
 :  Bioss
undefined
Anti-PRKD3 Rabbit Polyclonal Antibody
  BOSSBS-5564R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Protein kinase D3
  • Antigen symbol:
    PRKD3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Antigen synonyms:
    nu type|nPKC-NU|PKC-NU|PRKCN|nPKC-nu|Protein kinase C|PKD3|EPK2|Protein kinase EPK2
  • Modification:
    phospho-Ser41
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

Protein kinase C (PKC) is a family of serine and threonine specific protein kinases that can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role. The protein encoded by this gene is one of the PKC family members. This kinase can be activated rapidly by the agonists of G protein coupled receptors. It resides in both cytoplasm and nucleus, and its nuclear accumulation is found to be dramatically enhanced in response to its activation. This kinase can also be activated after B cell antigen receptor (BCR) engagement, which requires intact phopholipase C gamma and the involvement of other PKC family members.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: PRKD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse