Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase C Zeta
- Antigen symbol:PRKCZ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:5590
- Antigen synonyms:PRKCE|Protein kinase C beta type|PKCG|PKCE|PRKCT|PRKCG|SDK1|PKC2|PKC-A|nPKC-zeta|nPKC-theta|Protein kinase C zeta type|Protein kinase C epsilon type|PRKACA|nPKC-epsilon|PKC-B|PRKCZ|Protein kinase C alpha type|nPKC-delta|PKC-alpha|PRKCA|PKC-beta|PRKCQ|PKCB|PKC-gamma|Protein kinase C theta type|Tyrosine-protein kinase PRKCD|Protein kinase C gamma type|Sphingosine-dependent protein kinase-1|Protein kinase C delta type|PRKCD|PRKCB|PKC zeta|PRKCB1|PKCA
- Modification:Thr410
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Protein kinase C (PKC) zeta is a member of the PKC family of serine/threonine kinases which are involved in a variety of cellular processes such as proliferation, differentiation and secretion. Unlike the classical PKC isoenzymes which are calcium-dependent, PKC zeta exhibits a kinase activity which is independent of calcium and diacylglycerol but not of phosphatidylserine. Furthermore, it is insensitive to typical PKC inhibitors and cannot be activated by phorbol ester. Unlike the classical PKC isoenzymes, it has only a single zinc finger module. These structural and biochemical properties indicate that the zeta subspecies is related to, but distinct from other isoenzymes of PKC. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Jul 2008].
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: PRKCZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat