- Antibody type:Primary
- Antigen name:3-phosphoinositide dependent Protein kinase-1
- Antigen symbol:PDPK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:5170
- Antigen synonyms:3 Phosphoinositide Dependent Protein Kinase 1|PkB kinase|PkB kinase like gene 1|PDPK1|PDK 1|hPDK 1|PDPK1_HUMAN.|PkB like 1
- Modification:Ser396
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
PDK1 (3 Phosphoinositide Dependent Protein Kinase 1) phosphorylates AGC kinases. PDK1 activates conventional PKC and PKC zeta through phosphorylation of critical threonine residues in the activation loop. PDK1 also phosphorylates Protein Kinase B (PKB) at threonine 308 in the presence of phosphatidylinositol-3,4,5-trisphosphate. Active Akt inactivates Glycogen Synthase Kinase 3 (GSK3), eventually leading to the dephosphorylation and activation of glycogen synthase and the stimulation of glycogen synthesis. Because of the role that PDK plays in insulin-induced glycogen synthesis and PKC activation it is a potentially important target for metabolic drug research. There are three named isoforms.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: PDPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat