Anti-MYC Rabbit Polyclonal Antibody
Catalog # BOSSBS-4963R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:V-myc Avian Myelocytomatosis Viral Oncogene homolog
- Antigen symbol:MYC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Gene ID:4609
- Antigen synonyms:bHLHe39|Myc2|Nird|AU016757|Niard
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Transcription factor that binds DNA in a non-specific manner, yet also specifically recognizes the core sequence 5'-CAC[GA]TG-3'. Activates the transcription of growth-related genes.
IHC-P(1:100-500), IF(IHC-P)(1:50-200), IF(ICC)(1:50-200)
Type: Primary
Antigen: MYC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat