Anti-TAF1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-2812R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 250kDa
- Antigen symbol:TAF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Gene ID:6872
- Antigen synonyms:DYT3|TAF2A|DYT3/TAF1|P250|BA2R|CCGS|N-TAF1|NSCL2|KAT4|TAFII-250|CCG1|OF|TAFII250|TAF(II)250|XDP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Largest component and core scaffold of the TFIID basal transcription factor complex. Contains novel N- and C-terminal Ser/Thr kinase domains which can autophosphorylate or transphosphorylate other transcription factors. Phosphorylates TP53 on 'Thr-55' which leads to MDM2-mediated degradation of TP53. Phosphorylates GTF2A1 and GTF2F1 on Ser residues. Possesses DNA-binding activity. Essential for progression of the G1 phase of the cell cycle.
IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: TAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat