Specifications
- Antibody type:Primary
- Antigen name:Interleukin 16
- Antigen symbol:IL16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:3603
- Antigen synonyms:NIL16|Lymphocyte chemoattractant factor|prIL-16|IL-16|PRIL16|Pro-interleukin-16|LCF
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
IL16 is a proinflammatory cytokine that is chemotactic for CD4+ T lymphocytes, monocytes, and eosinophils. IL16 is synthesized as a precursor molecule (pro IL16) of approximately 68 kDa and 631 amino acid residues lacking a signal peptide. The sequence and structure of IL16 is conserved across species. In addition to inducing chemotaxis, IL16 upregulates the IL2 receptor and upregulates HLADR expression. It also inhibits T cell receptor (TCR)/CD3 dependent activation, and suppresses HIV1 replication in vitro. Sources of IL16 include epithelial cells, mast cells, T lymphocytes (CD4+ and CD8+), macrophages, synovial fibroblasts, and eosinophils.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: IL16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat