Order Entry
Austria
ContactUsLinkComponent
Anti-SP-A Rabbit Polyclonal Antibody
Anti-SP-A Rabbit Polyclonal Antibody
Catalog # BOSSBS-10265R
Supplier:  Bioss
Anti-SP-A Rabbit Polyclonal Antibody
Catalog # BOSSBS-10265R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Surfactant protein A
  • Antigen symbol:
    SP-A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Antigen synonyms:
    Surfactant Protein A|Pulmonary surfactant associated protein|SFTPA1B|pulmonary surfactant-associated glycoprotein A|SP-A2|SFPA2_MOUSE|Pulmonary surfactant-associated protein A2 precursor|SFTPA1|SP A|SP-A|SFTP1|PSP-A|Alveolar proteinosis protein|SFTPA|COLEC4|Pulmonary surfactant apoprotein|Surfactant pulmonary associated protein A1|Collectin-5.|PSPA|35 kDa pulmonary surfactant-associated protein|MGC133365|SP A1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

This gene is one of several genes encoding pulmonary-surfactant associated proteins (SFTPA) located on chromosome 10. Mutations in this gene and a highly similar gene located nearby, which affect the highly conserved carbohydrate recognition domain, are associated with idiopathic pulmonary fibrosis. The current version of the assembly displays only a single centromeric SFTPA gene pair rather than the two gene pairs shown in the previous assembly which were thought to have resulted from a duplication. [provided by RefSeq, Sep 2009].

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SP-A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat