Orders LinkContactUsLinkComponent
Robotic tips, Matrix
Robotic tips, Matrix
30 µl, Black, FLIPR Liberty Type B, 10 racks, 384 each
Catalog # MATR5361
   Low Manufacturing Impact
Robotic tips, Matrix
Catalog # MATR5361
   Low Manufacturing Impact
Robotic tips, Matrix
30 µl, Black, FLIPR Liberty Type B, 10 racks, 384 each
Price
1 067,00 €
Log in for availability
1 * 3.840 ST
Product Details & Documents
These automation tips are compatible with 384 format Beckman instruments, PerkinElmer and FLIPR instruments using CCS Type A pipetting heads and FLIPR Type B Liberty systems. All of these tips are validated on their compatible instruments to ensure the highest quality for pipetting in automated laboratories.
  • Performance validation on compatible instruments includes 1536 target straightness testing, leak testing, length, weight and wall thickness testing, and pipetting head seal force testing
  • Design includes molded conductive rack and snap plate to hold tips to reduce static interactions during shipping and seating on instrument pipetting heads, enhancing product performance and reliability
Specifications
  • Packed:
    10 racks, 384 each
  • Volume:
    30 µl
  • Tip colour:
    Black
  • For use with:
    FLIPR Liberty Type B
  • Cat. No.:
    MATR5361
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact