Assessment cabinets, Colorbox
The Colorbox is used to provide the illumination conditions for any visual inspection at a 45° angle.
- Both interior and exterior finishes according to high quality standard
- Multiple selectable light sources
- Assessments of gloss, structure, damages and metamerism
: Supplied with removable viewing table and counter.
- Catalog No:
- SIMEVF1200
- SIMEVF0600
- Angle:
- 45°
Frequently Bought Together