Tubing, copper
Supplier: Agilent
Certificates
About this item
Copper, precleaned.
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Screw caps for certified screw top vials
Supplier: Agilent
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection