Specifications
- W×D×H ext.:1245×615×1285 mm
- Treated air volume:200−230
- Description:Captair® 481 Smart, EU-plug
- Model:Captair® Smart 481
- Air velocity at the front panel:0,4 - 0,6 m/s
- Filters:HEPA H14: filtration efficiency: 99,995 % according to MPPS methods, EN1822 standard
ULPA U17: filtration efficiency 99,999995 % according to MPPS method, EN 1822 standard - Lighting:LED > 650 lux
- Power:60 W
- Pk:1
- W×D×H int.:1176×533×1100 mm
Specifications
About this item
The new Captair® Smart ductless filtering hoods are designed with a simple and innovative way of communication. Smart Technology, simple and intuitive communication by light indicates the status of the hood. A simple light pattern will indicate if the airflow is compromised, if there is a fan failure or in case of a filter breakthrough.
Please complete the quote request to enable us to assist you in finding the right unit for your application.
https://uk.cmd2.vwr.com/bin/public/fc/main?f=Y5&pimid=17327608
- The Erlab's unique Flex Filtration can manage most laboratory applications
- New sensors are added for detection of solvents or acids or formalehyde
- Low power consumption
- New electronic anemometer monitors face velocity
- The 6° slanted sash offers optimal visibility and ergonomic work space
- Large openings for easy access to the work
The configurable filtering system above the enclosure can be adapted to filter gases, solvents, powders and particulates keeping the user and the laboratory protected by safely and efficiently recirculating air within the room and releasing purified air back into the room free of any toxic hazardous vapours or odours.
The monitoring gives the user the control to monitor the hood, change the settings and delivers safety alerts immediately to the mobile, tablet or PC device.
The structure is made from corrosion resistant electro-galvanised steel coated with anti-acid polymer, sides and front panels are in chemical resistant acrylic.
Types of carbon filters:
AS: For organics vapours
BE: Polyvalent for acid and organic vapours
F: For formaldehyde vapours
K: For ammonia vapours
HP: H14 HEPA filter for powders
Filtration fault detection: Detection sensors for solvents, acids or formaldehyde.
Monitoring for ambient handling conditions: Temperature & Hygrometry sensors.