Filter tips, ART® Ergonomic
Supplier: Thermo Fisher Scientific
Certificates
About this item
The unique bi-fabricated wall of the ART® Ergonomic tip creates a strong, effective seal while requiring significantly lower tip insertion and tip ejection force. Available for Gilson Pipetman and FinnPipette Focus + Digital pipettes.
- Reduce strains and injury
- Secure and consistent seals
- Less ejection force
- Available in low retention polymer
Limited Time Sale
Select items below are currently on sale
Specifications
- Catalog No:
- 736-0210
- 736-0208
- 613-3865
- 613-3866
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)