Order Entry
Austria
ContactUsLinkComponent
VWR®, Buffer, ammonia solution, pH 10 ready to use
VWR®, Buffer, ammonia solution, pH 10 ready to use
Catalog # 160204A
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Buffer, ammonia solution, pH 10 ready to use
Catalog # 160204A
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    Buffer, ammonia solution for determination of hardness of water
  • Seq:
    01
  • Contents:
    Ammonia w/v: 14,5 - 15,5%

Specifications

About this item

Ideal for hardness of water determination.