Orders LinkContactUsLinkComponent
Calibration weight sets, class E2, Proof Line
Calibration weight sets, class E2, Proof Line
Supplier:   Sartorius Balances
Calibration weight sets, class E2, Proof Line
Supplier:  Sartorius Balances
Certificates
Calibration weight sets, class E2, Proof Line
Calibration weight sets, class E2, Proof Line
9 Options Available Below
Product Details & Documents
These class E2 weight sets include aluminium (1 to 5 mg) or nickel silver (10 to 500 mg) or stainless steel (1 g and higher) weights.
Product Family Options
E2 Weight sets without certificate
Product Information
  • Weight set
    Type
    Availability
    Price
  • 1 mg - 500 mg
    Without certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 531,00 €
    531,00 €
    1 * 1 ST
       531,00 €
  • 1 g - 100 g
    Without certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 130,00 €
    1 130,00 €
    1 * 1 ST
       1 130,00 €
  • 1 g - 200 g
    Without certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 350,00 €
    1 350,00 €
    1 * 1 ST
       1 350,00 €
  • 1 g - 1 kg
    Without certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 160,00 €
    2 160,00 €
    1 * 1 ST
       2 160,00 €
  • 1 g - 5 kg
    Without certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 270,00 €
    4 270,00 €
    1 * 1 ST
       4 270,00 €
E2 Weight sets with DAkkS certificate
Product Information
  • Weight set
    Type
    Availability
    Price
  • 1 mg - 500 mg
    With DAkkS certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 957,00 €
    957,00 €
    1 * 1 ST
       957,00 €
  • 1 g - 200 g
    With DAkkS certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 740,00 €
    1 740,00 €
    1 * 1 ST
       1 740,00 €
  • 1 g - 1 kg
    With DAkkS certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 600,00 €
    2 600,00 €
    1 * 1 ST
       2 600,00 €
  • 1 g - 5 kg
    With DAkkS certificate
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 910,00 €
    4 910,00 €
    1 * 1 ST
       4 910,00 €