Orders LinkContactUsLinkComponent
Precision hydrometers, ASTM (API Grade)
Precision hydrometers, ASTM (API Grade)
Precision hydrometers, ASTM (API Grade)
Precision hydrometers, ASTM (API Grade)
These precision hydrometers are calibrated without thermometer.
Error limit: ±0,1° API
Reference temperature: 15,5 °C
 : American Petroleum Institute (API) and ASTM E100-10.
 : Each packed in a square plastic sleeve.
Product Family Options
  • Description
    Model
    Range
    Division
    Length
  • ASTM hydrometer
    1H-62
    −1…+11 °API
    0,1 °API
    330 mm
    1 * 1 ST
    1 * 1 ST - 36,00 €
    36,00 €
    1 * 1 ST
       36,00 €
  • ASTM hydrometer
    2H-62
    9…21 °API
    0,1 °API
    330 mm
    1 * 1 ST
    1 * 1 ST - 36,00 €
    36,00 €
    1 * 1 ST
       36,00 €
  • ASTM hydrometer
    3H-62
    19…31 °API
    0,1 °API
    330 mm
    1 * 1 ST
    1 * 1 ST - 36,00 €
    36,00 €
    1 * 1 ST
       36,00 €
  • ASTM hydrometer
    4H-62
    29…41 °API
    0,1 °API
    330 mm
    1 * 1 ST
    1 * 1 ST - 36,00 €
    36,00 €
    1 * 1 ST
       36,00 €
  • ASTM hydrometer
    5H-62
    39…51 °API
    0,1 °API
    330 mm
    1 * 1 ST
    1 * 1 ST - 36,00 €
    36,00 €
    1 * 1 ST
       36,00 €
  • ASTM hydrometer
    6H-62
    49…61 °API
    0,1 °API
    330 mm
    1 * 1 ST
    1 * 1 ST - 36,00 €
    36,00 €
    1 * 1 ST
       36,00 €
  • ASTM hydrometer
    7H-62
    59…71 °API
    0,1 °API
    330 mm
    1 * 1 ST
    1 * 1 ST - 36,00 €
    36,00 €
    1 * 1 ST
       36,00 €
  • ASTM hydrometer
    8H-62
    69…81 °API
    0,1 °API
    330 mm
    1 * 1 ST
    1 * 1 ST - 39,30 €
    39,30 €
    1 * 1 ST
       39,30 €
  • ASTM hydrometer
    9H-62
    79…91 °API
    0,1 °API
    330 mm
    1 * 1 ST
    1 * 1 ST - 39,30 €
    39,30 €
    1 * 1 ST
       39,30 €
  • ASTM hydrometer
    10H-62
    89…101 °API
    0,1 °API
    330 mm
    1 * 1 ST
    1 * 1 ST - 39,30 €
    39,30 €
    1 * 1 ST
       39,30 €