Order Entry
Austria
Orders LinkContactUsLinkComponent
VWR®, pH buffer capsules

VWR®, pH buffer capsules

VWR®, pH buffer capsules
Supplier:  VWR Chemicals
Certificates
VWR®, pH buffer capsules
VWR®, pH buffer capsules

2 Options Available Below

Product Details & Documents
  • Colour coded for easy identification
  • Preservative-free
  • Extended shelf life
  • Add one capsule to 100 ml of distilled water
Product Family Options

Product Information

  • pH value
    pH accuracy
    Colour
    Final volume
    Pack type
    Pk Info
    Pk
    Availability
    Price
  • Capsules
    4,01
    ±0,02
    Orange
    100 ml
    Plastic tube
    5 plastic tubes, 10 capsules/tube
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 228,00 €
    228,00 €
    1 * 50 ST
       228,00 €

  • Capsules
    7,00
    ±0,02
    Green
    100 ml
    Plastic tube
    5 plastic tubes, 10 capsules/tube
    50
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 228,00 €
    228,00 €
    1 * 50 ST
       228,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.