Anti-FXYD1 Rabbit Polyclonal Antibody
Catalog # BSBTPB9704
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:FXYD domain containing ion transport regulator 1
- Antigen symbol:FXYD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5348
- Antigen synonyms:PLM
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Phospholemman(FXYD1) detection. Tested with WB, IHC-P in Human;Rat;Mouse.
Type: Primary
Antigen: FXYD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat