Orders LinkContactUsLinkComponent
API precision hydrometers
API precision hydrometers
Supplier:   Ludwig Schneider
API precision hydrometers
Supplier:  Ludwig Schneider
Certificates
API precision hydrometers
API precision hydrometers
10 Options Available Below
Product Details & Documents
API precision hydrometers with blue filling thermometer.
Reference temperature: 15,5 °C
Error limit: ±0,1 °API
Zertifizierung: ASTM E2995-14.
Bestellinformation: A works or DAkkS calibration certificate is available (on request).
Verpackung: Each packed in a square plastic sleeve.
Product Family Options
Product Information
  • Model
    Range
    Division
    Temperature Range
    Length
    Availability
    Price
  • S551 HL-14
    −1…+11 °API
    0,1 °API
    −18…+66 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,20 €
    84,20 €
    1 * 1 ST
       84,20 €
  • S552 HL-14
    9…21 °API
    0,1 °API
    −18…+66 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,20 €
    84,20 €
    1 * 1 ST
       84,20 €
  • S553 HL-14
    19…31 °API
    0,1 °API
    −18…+66 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,20 €
    84,20 €
    1 * 1 ST
       84,20 €
  • S554 HL-14
    29…41 °API
    0,1 °API
    −18…+66 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,20 €
    84,20 €
    1 * 1 ST
       84,20 €
  • S555 HL-14
    39…51 °API
    0,1 °API
    −18…+66 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,20 €
    84,20 €
    1 * 1 ST
       84,20 €
  • S556 HL-14
    49…61 °API
    0,1 °API
    −18…+66 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,20 €
    84,20 €
    1 * 1 ST
       84,20 €
  • S557 HL-14
    59…71 °API
    0,1 °API
    −18…+66 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,20 €
    84,20 €
    1 * 1 ST
       84,20 €
  • S558 HL-14
    69…81 °API
    0,1 °API
    −18…+66 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,20 €
    84,20 €
    1 * 1 ST
       84,20 €
  • S559 HL-14
    79…91 °API
    0,1 °API
    −18…+66 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,20 €
    84,20 €
    1 * 1 ST
       84,20 €
  • S560 HL-14
    89…101 °API
    0,1 °API
    −18…+66 °C
    380 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,20 €
    84,20 €
    1 * 1 ST
       84,20 €