ContactUsLinkComponent
Flexible skeleton model - Fred
Flexible skeleton model - Fred
Catalog # BINH1013873
Supplier:  3B Scientific
undefined
Flexible skeleton model - Fred
Catalog # BINH1013873
Supplier:  3B Scientific
Specifications
  • Description:
    Flexible skeleton model - Fred
  • Pk:
    1
Specifications
About this item
Fred offers all the advantages of a skeleton. Fred's bendy spine can copy almost any human movement too. Once bent, Fred will remain in that position, to show correct and incorrect postures as well as any pathological malalignments. All skull movements can be shown on the head joints. Protruding spinal nerves and vertebral arteries are also shown on this skeleton, as well as a dorsolateral slipped disc between the 3rd and 4th lumbar vertebrae.
  • Top quality, life-size natural casting 'Made in Germany'
  • Made from a durable, unbreakable synthetic material
  • 3-part assembled skull with individually inserted teeth
  • Close to the realistic weight of around 200 bones
  • On a stable metal stand with 5 casters