Order Entry
Austria
ContactUsLinkComponent
Steel safe for radioactive materials
Steel safe for radioactive materials
Catalog # BINHU8483219
Supplier:  3B Scientific
undefined
Steel safe for radioactive materials
Catalog # BINHU8483219
Supplier:  3B Scientific

Specifications

  • Description:
    Steel safe for theft proof storage of radioactive materials in accordance with radiation protection requirements
  • W×D×H:
    300×140×360 mm
  • Doors:
    1
  • Pk:
    1
  • Weight:
    3 kg

Specifications

About this item