ContactUsLinkComponent
SPE flow filters, Resprep®
SPE flow filters, Resprep®
Catalog # REST26024
Supplier:  Restek
undefined
SPE flow filters, Resprep®
Catalog # REST26024
Supplier:  Restek
Specifications
  • Description:
    Flow filters
  • Pk:
    20
Specifications
About this item
Flow filters are designed to improve flow when filtering oil and grease samples. These filters can be used with Resprep® Diskcover-47 reservoir or any 47 mm glass sample reservoir.