Orders LinkContactUsLinkComponent
Solid phase extraction disks, Resprep® Resin
Solid phase extraction disks, Resprep® Resin
Solid phase extraction disks, Resprep® Resin, 47 mm
Catalog # REST26023
Supplier:   Restek
Solid phase extraction disks, Resprep® Resin
Catalog # REST26023
Supplier:  Restek
Solid phase extraction disks, Resprep® Resin
Solid phase extraction disks, Resprep® Resin
Solid phase extraction disks, Resprep® Resin, 47 mm
Price
450,19 €
Log in for availability
1 * 20 ST
Product Details & Documents
Glass fiber disks embedded with styrene/DVB resin. The disks are used for chlorinated, benzidine-containing or nitrogen-containing pesticides.
 : Meets the requirements of EPA methods 515.2 and 553.
Specifications
  • Ø:
    47 mm
  • Description:
    Solid phase extraction disks, Resprep® Resin
  • Seq:
    01