Vacuum/pressure station, oil-free
Supplier: Restek
Certificates
About this item
Vacuum/pressure station allows switching from pressure to vacuum in a matter of seconds. It is quiet and has a oil-free unit which will not contaminate the extraction system or samples. Provides 20" Hg (68 kPa) vacuum or 18 psi (124 kPa) pressure.
Frequently Bought Together