ContactUsLinkComponent
MSD source nut
MSD source nut
Catalog # REST20643
Supplier:  Restek
undefined
MSD source nut
Catalog # REST20643
Supplier:  Restek
Specifications
  • Description:
    Detector MSD source nut
  • Material:
    Brass
  • Pk:
    2
Specifications
About this item
This MSD source nut is made from brass. The 1,2 mm nut bore permits easy removal of ferrules with a standard tapered needle file. The material prevent thread-stripping on the transfer line, the design enhances ease of threading onto the transfer line and improves overall lifetime.