Solid phase extraction disks, Resprep®
Supplier: Restek
Jump to:
About this item
These glass fiber disks are embedded with C18 or C8 bonded silica. Deep-pore design reduces clogging and allows faster flow rates. Semivolatile organic compounds can be etracted.
: Meet the requirements for EPA methods 525.1, 506, 550.1 and 549.1.
3 Options Available Below
Product Details & Documents
These glass fiber disks are embedded with C18 or C8 bonded silica. Deep-pore design reduces clogging and allows faster flow rates. Semivolatile organic compounds can be etracted.
: Meet the requirements for EPA methods 525.1, 506, 550.1 and 549.1.